AuftragsverfolgungKontakt
Inlet liners | Avantor
Inlet liners
O-ring for high temperature PTV
Artikelnr. HEWL5188-5311
Anbieter:   Agilent
Inlet liners
Artikelnr. HEWL5188-5311
Anbieter:  Agilent
Inlet liners
Inlet liners
O-ring for high temperature PTV
Preis
156,00 €
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
These inlet liners prevent adsorption and ensure accurate sample transfer onto the GC column through a robust deactivated surface. These liners are recommended for trace level analysis of active analytes. Liners with inert wool retain non-volatiles, prolonging column life and lowering frequency of MS source maintenance.
  • Consistent high level of inertness
Spezifikationen
  • Bezeichnung:
    O-ring for high temperature PTV
  • Seq:
    55