INSOLUTION(TM) EPOXOMICIN SYNTHETIC 1 * 50 µG
Artikelnr. 324801-50
Lieferant: Merck Millipore (Calbiochem)
INSOLUTION(TM) EPOXOMICIN SYNTHETIC 1 * 50 µG
Artikelnr. 324801-50
Lieferant: Merck Millipore (Calbiochem)
Frequently Bought Together
VWR®, Serologische Pipetten, Standard Serie
Lieferant: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Lieferant: VWR Collection
VWR®, PCR-Gefäße, 0,2 ml
Lieferant: VWR Collection
Aceton ≥99%, TECHNICAL
Lieferant: VWR Chemicals
VWR®, Transferpipetten, Standard Line
Lieferant: VWR Collection
VWR®, Nitrilhandschuhe
Lieferant: VWR Collection
VWR®, Transferpipetten, High Performance
Lieferant: VWR Collection
VWR®, Spreaders, L-shaped, Disposable
Lieferant: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
Lieferant: EPPENDORF
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Lieferant: EPPENDORF
Safe-Lock Tubes, 2,0 ml - Microtubes
Lieferant: EPPENDORF
Mammalian protein extract reagents, M-PER™
Lieferant: Thermo Fisher Scientific