Capillary nuts
Enhanced capillary nut
Artikelnr. REST20375
Anbieter: Restek
Springe zu:
Product Details & Documents
These capillary nuts are ideal for Shimadzu 17A, 2010, and 2014 GCs.
The enhanced capillary nut design eliminates the slot, increasing lifetime and durability.
Spezifikationen
- Bezeichnung:Enhanced capillary nut
- Seq:02
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.