Precision sampling valves for vials, Mininert®
Mininert® precision sampling valves, for crimp-top vials
Artikelnr. REST24905
Anbieter: Restek
Springe zu:
Product Details & Documents
Mininert® valves are very convenient for repetitive sampling and limit content exposure to the silicon septum. The crimp-top valve for 20 mm ID glassware slides into the neck of the vial.
- Available for screw cap and crimp top vials
- Threaded flange used to secure tight fit
Spezifikationen
- Bezeichnung:Mininert® precision sampling valves, for crimp-top vials
- Seq:06