Flow controllers, Veriflo
4 – 8 ml/min, Siltek® behandeltes Edelstahl, Flow controller, 0,041 mm
Artikelnr. REST24256
Anbieter: Restek
Springe zu:
Product Details & Documents
Veriflo controllers can be used with passive air samplers in maintaining a constant flow rate.
Entire sample flow path is made of stainless steel or siltek® treated for maximum inertness.
Spezifikationen
- Ø:0,041 mm
- Maximale Durchflussrate:4 – 8 ml/min
- Bezeichnung:Flow controller
- Material:Siltek® behandeltes Edelstahl
- Seq:08