Flow controllers, Veriflo
0,5 – 2 ml/min, Edelstahl, Replacement orifice, 0,02 mm
Artikelnr. REST24218
Anbieter: Restek
Springe zu:
Product Details & Documents
Veriflo controllers can be used with passive air samplers in maintaining a constant flow rate.
Entire sample flow path is made of stainless steel or siltek® treated for maximum inertness.
Spezifikationen
- Ø:0,02 mm
- Maximale Durchflussrate:0,5 – 2 ml/min
- Bezeichnung:Replacement orifice
- Material:Edelstahl
- Seq:101