Solid phase extraction cartridges, Resprep®
SPE Florisil cartridges, Kartusche, 6 ml, 1000 mg
Catalog # REST24034
Supplier: Restek
Springe zu:
Product Details & Documents
These cartridges are manufactured using high density polypropylene and have polyethylene frits unless otherwise noted. Hydrophilic (polar) adsorbents used to extract hydrophilic analytes from nonpolar matrices, such as organic solvents (e.g., polar contaminants from sample extracts).
Cartridges may be processed by any one or all of these techniques: positive pressure, sidearm flask, centrifuge, or vacuum manifold.
Spezifikationen
- Säulenform:Kartusche
- Kapazität:6 ml
- Bezeichnung:SPE Florisil cartridges
- Seq:03
- Bettgewicht:1000 mg