Rethreading tool
Rethreading tool, for 1/8" compression fitting
Artikelnr. REST23017
Anbieter: Restek
Springe zu:
Product Details & Documents
These rethreading tools are suitable for repairing worn or damaged threads that come as a result of consistent use and temperature changes.
- Multiple uses (injection ports, fittings, etc.)
- Built-in guide prevents cross-threading
Repeated installation, removal and temperature changes cause wear and damage to threads on GC parts. This can cause a poor seal, and oxygen can enter the system, compromising analytical results and possibly destroying expensive analytical columns.
Spezifikationen
- Bezeichnung:Rethreading tool, for 1/8" compression fitting
- Seq:01