Capillary column nuts for Agilent GCs
Capillary column nut, for use with standard 1/16" type ferrules, Messing
REST21879
: Restek
These nuts are designed for Agilent 5890/6850/6890/7890 GCs. Nuts for use with standard ¹/₁₆" type ferrules are designed to fit a wider variety of ¹/₁₆" ferrules.
- Bezeichnung:Capillary column nut, for use with standard 1/16" type ferrules
- Material:Messing
- Seq:04