AuftragsverfolgungKontakt
Capillary column nuts for Agilent GCs
Capillary column nut, for use with standard 1/16" type ferrules, Messing
  REST21879
 :   Restek
Capillary column nuts for Agilent GCs
  REST21879
 :  Restek
Capillary column nuts for Agilent GCs
Capillary column nuts for Agilent GCs
Capillary column nut, for use with standard 1/16" type ferrules, Messing
56,67 €
1 * 2 ST
These nuts are designed for Agilent 5890/6850/6890/7890 GCs. Nuts for use with standard ¹/₁₆" type ferrules are designed to fit a wider variety of ¹/₁₆" ferrules.
  • Bezeichnung:
    Capillary column nut, for use with standard 1/16" type ferrules
  • Material:
    Messing
  • Seq:
    04