Express Eingabe
Austria
AuftragsverfolgungKontakt
O-rings, graphite | Avantor

O-rings, graphite

Graphite O-rings, for liners with outer Ø 4 mm
Artikelnr. REST21476
Anbieter:   Restek
O-rings, graphite
Artikelnr. REST21476
Anbieter:  Restek
O-rings, graphite
O-rings, graphite

Graphite O-rings, for liners with outer Ø 4 mm

Preis
Preis anfragen
Einloggen für Verfügbarkeit
1 * 25 ST
Product Details & Documents

Graphite O-rings for PerkinElmer Auto SYS XL or Clarus GCs with PSS injector.

Max temp.: 450 °C

Spezifikationen
  • Bezeichnung:
    Graphite O-rings, for liners with outer Ø 4 mm
  • Seq:
    02
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.