O-rings, graphite
Graphite O-rings, for liners with outer Ø 4 mm
REST21475
: Restek
Graphite O-rings for PerkinElmer Auto SYS XL or Clarus GCs with PSS injector.
Max temp.: 450 °C
- Bezeichnung:Graphite O-rings, for liners with outer Ø 4 mm
- Seq:01
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



