Capillary installation gauges
Capillary installation gauge for 1/16" fittings (1/16" ferrules)
Artikelnr. REST21399
Anbieter: Restek
Springe zu:
Product Details & Documents
These capillary installation gauges for Agilent GCs are made from high-quality stainless steel.
- Prevents crushed column ends
- Seats graphite ferrules onto column for consistent installations
Spezifikationen
- Bezeichnung:Capillary installation gauge for 1/16" fittings (1/16" ferrules)
- Seq:02