Express Eingabe
Austria
AuftragsverfolgungKontakt
Packed GC columns, 2abc refinery column set | Avantor
Packed GC columns, 2abc refinery column set
Analytical controls configuration
Artikelnr. REST88000-875
Anbieter:   Restek
Packed GC columns, 2abc refinery column set
Artikelnr. REST88000-875
Anbieter:  Restek
Packed GC columns, 2abc refinery column set
Analytical controls configuration
Preis
2 371,60 €
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
Special columns for unsaturated light hydrocarbons.
  • Faster separations of C1 to C4 hydrocarbons
When used in the proper valving system, it will elute C5+ hydrocarbons ahead of C1 through C4 hydrocarbons.
Spezifikationen
  • Bezeichnung:
    Analytical controls configuration
  • Seq:
    02
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.