Packed GC columns, 2abc refinery column set
Analytical controls configuration
Artikelnr. REST88000-875
Anbieter: Restek
Springe zu:
Product Details & Documents
Special columns for unsaturated light hydrocarbons.
- Faster separations of C1 to C4 hydrocarbons
When used in the proper valving system, it will elute C5+ hydrocarbons ahead of C1 through C4 hydrocarbons.
Spezifikationen
- Bezeichnung:Analytical controls configuration
- Seq:02