Hoffmann pinch cocks, Rotilabo®
13 mm
Artikelnr. ROTH1585.1
Marketsource
Anbieter: Roth Carl
Springe zu:
Product Details & Documents
Rotilabo®-tube clamping screws made of chrome-plated metal for fine regulation.
- Tube clamping screws according to Hoffmann
- Hinged bottom bar can be swing open
Spezifikationen
- Seq:01
- Bandbreite:13 mm