Kolophonium
5 kg
Artikelnr. ROTH9051.2
Marketsource
Anbieter: Roth Carl
Springe zu:
Product Details & Documents
Synonyme: Rosin, gum, Rosin, Gum rosin
- CAS Nummer:8050-09-7
- MDL-Nummer:MFCD00132205
- Schmelzpunkt:75…81 °C
- Dichte:1,1 g/cm³ (20 °C)
- Blitzpunkt:188 °C
- EINECS:232-475-7
Spezifikationen
Spezifikationen
- VE:5 kg
Spezifikationen
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



