Express Eingabe
Austria
AuftragsverfolgungKontakt
 

Polyethylenglycol 2.000

5 kg
Catalog # ROTH0154.2
Marketsource
Supplier:   Roth Carl
Polyethylenglycol 2.000
Catalog # ROTH0154.2
Supplier:  Roth Carl
Marketsource
CAS Number:  25322-68-3
Polyethylenglycol 2.000

5 kg

CAS Number:  25322-68-3
Price
338,00 €
Log in for availability
1 * 5 kg
Product Details & Documents
Synonyms:  PEG 2000, Ethylene glycol polymer 2000, PEG 2,000, Ethylene glycol polymer average Mn 2000
  • CAS Number:25322-68-3
  • Formula:H(OCH₂CH₂)nOH
  • MDL Number:MFCD00081839
  • Molecular Weight:2000 g/mol
  • Melting Pt:49…52 °C
  • Density:1,21 g/cm³ (20 °C)
  • Flash Pt:250 °C
  • Storage Temperature:Raumtemperatur
  • EINECS:500-038-2
Specifications

Specifications

  • VE:
    5 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.