Express Eingabe
Austria
AuftragsverfolgungKontakt
Quecksilber

Quecksilber

Glass bottle for solids, 1 kg
Catalog # ROTH7593.2
Toxic
Health
Environment
Marketsource
Supplier:   Roth Carl
Quecksilber
Catalog # ROTH7593.2
Supplier:  Roth Carl
Marketsource
CAS Number:  7439-97-6
Quecksilber

Glass bottle for solids, 1 kg

CAS Number:  7439-97-6
Price
280,00 €
Log in for availability
1 * 1 kg
Product Details & Documents
  • CAS Number:7439-97-6
  • Formula:Hg
  • MDL Number:MFCD00011035
  • Molecular Weight:200.59 g/mol
  • Merck Index:13,05925
  • Boiling Pt:356,5 °C (1013 hPa)
  • Melting Pt:–39 °C
  • Density:13,54 g/cm³ (20 °C)
  • Storage Temperature:Raumtemperatur
  • EINECS:231-106-7
  • UN:2809
  • ADR:8,III
Specifications

Specifications

  • Packung:
    Glass bottle for solids
  • VE:
    1 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.