Express Eingabe
Austria
AuftragsverfolgungKontakt
 

AMPSO

Glasflasche für Feststoffe, 25 g
Catalog # ROTH7159.1
Marketsource
Supplier:   Roth Carl
AMPSO
Catalog # ROTH7159.1
Supplier:  Roth Carl
Marketsource
CAS Number:  68399-79-1
AMPSO

Glasflasche für Feststoffe, 25 g

CAS Number:  68399-79-1
Price
65,30 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  N-(1,1-Dimethyl-2-hydroxyethyl)-3-amino-2-hydroxypropanesulphonic acid
  • CAS Number:68399-79-1
  • Formula:C₇H₁₇NO₅S
  • MDL Number:MFCD00041777
  • Molecular Weight:227.28 g/mol
  • Storage Temperature:Raumtemperatur
  • EINECS:269-991-7
Specifications

Specifications

  • Packung:
    Glasflasche für Feststoffe
  • VE:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.