Essigsäure
Glasflasche, 2,5 L
Artikelnr. ROTH3738.5
Marketsource
Anbieter: Roth Carl
Springe zu:
Product Details & Documents
Synonyme: Vinegar acid, Methanecarboxylic acid, Ethanoic acid
- CAS Nummer:64-19-7
- Formel:H₃CCOOH
- MDL-Nummer:MFCD00036152
- Molekulargewicht:60.05 g/mol
- Siedepunkt:118 °C (1013 hPa)
- Schmelzpunkt:17 °C
- Dichte:1,05 g/cm³ (20 °C)
- Blitzpunkt:38,5 °C
- Lagertemperatur:Raumtemperatur
- EINECS:200-580-7
- UN:2789
- ADR:8,II
- REACH:01-2119475328-30
Spezifikationen
Spezifikationen
- Packung:Glasflasche
- VE:2,5 L
Spezifikationen
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



