Graphite sealing rings
Graphite sealing ring, 2 pack
Catalog # REST21899
Supplier: Restek
Jump to:
Product Details & Documents
These graphite sealing rings are suitable for Thermo Scientific TRACE, 8000, 8000 TOP and Focus SSL instruments. They fit liners with 8 mm outer diameter.
Specifications
- Bezeichnung:Graphite sealing ring, 2 pack
- Seq:02
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.