Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
KIT SUREBLUE TMB 1-C MW (4X100ML) 1 * 1 KIT
Catalog # KPLI52-00-02
Supplier:
SERACARE LIFE SCIENCES
Certificates
Expand image
KIT SUREBLUE TMB 1-C MW (4X100ML) 1 * 1 KIT
Catalog # KPLI52-00-02
Supplier:
SERACARE LIFE SCIENCES
Certificates
Related Information
Documentation
Certificates
Search by Lot Number
Documentation
Frequently Bought Together
VWR® Inoculating Loops and Needles, Disposable
Supplier: VWR Collection
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
VWR®, Serologische Pipetten, Standard Serie
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
VWR®, Transferpipetten, High Performance
Supplier: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
VWR®, Spreaders, L-shaped, Disposable
Supplier: VWR Collection
VWR®, Geradwandige Probenbehälter, mit Schraubverschluss
Supplier: VWR Collection
VWR®, PCR-Gefäße, 0,2 ml
Supplier: VWR Collection
VWR®, Transferpipetten, Standard Line
Supplier: VWR Collection