Express Eingabe
Austria
AuftragsverfolgungKontakt
Glass wool
Deactivated borosilicate glass wool, more flexible than fused silica wool
Artikelnr. REST20789
Anbieter:   Restek
Glass wool
Artikelnr. REST20789
Anbieter:  Restek
Glass wool
Glass wool
Deactivated borosilicate glass wool, more flexible than fused silica wool
Preis
50,00 €
Einloggen für Verfügbarkeit
1 * 10 g
Product Details & Documents
Spezifikationen
  • Bezeichnung:
    Deactivated borosilicate glass wool, more flexible than fused silica wool
  • Seq:
    02
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.