Septum nuts
Septum nut, autosampler and PTV (for 23-gauge needles)
Artikelnr. REST20631
Anbieter: Restek
Springe zu:
Product Details & Documents
These septum nuts are made of high-quality stainless steel and for use with Agilent 5890/6850/6890/7890 split or splitless injectors.
- Needle guide allows easy penetration and prevents septum coring
- Manual injection septum nut allows use of 26-gauge needles for on-column injections
Spezifikationen
- Bezeichnung:Septum nut, autosampler and PTV (for 23-gauge needles)
- Seq:01