Express Eingabe
Austria
AuftragsverfolgungKontakt
Septum nuts | Avantor
Septum nuts
Septum nut, autosampler and PTV (for 23-gauge needles)
Artikelnr. REST20631
Anbieter:   Restek
Septum nuts
Artikelnr. REST20631
Anbieter:  Restek
Septum nuts
Septum nuts
Septum nut, autosampler and PTV (for 23-gauge needles)
Preis
69,26 €
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
These septum nuts are made of high-quality stainless steel and for use with Agilent 5890/6850/6890/7890 split or splitless injectors.
  • Needle guide allows easy penetration and prevents septum coring
  • Manual injection septum nut allows use of 26-gauge needles for on-column injections
Spezifikationen
  • Bezeichnung:
    Septum nut, autosampler and PTV (for 23-gauge needles)
  • Seq:
    01
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.