Express Eingabe
Austria
AuftragsverfolgungKontakt
O-rings, graphite | Avantor

O-rings, graphite

O-rings for splitless liners, I.Ø: 6,50 mm
Artikelnr. REST20298
Anbieter:   Restek
O-rings, graphite
Artikelnr. REST20298
Anbieter:  Restek
O-rings, graphite
O-rings, graphite

O-rings for splitless liners, I.Ø: 6,50 mm

Preis
77,65 €
Einloggen für Verfügbarkeit
1 * 10 ST
Product Details & Documents

These O-rings are made of graphite. Ideal for Bruker/Varian 1177 injectors.

  • Maximum operating temperature 450 °C
  • Can used with split or splitless liners
Spezifikationen
  • Bezeichnung:
    O-rings for splitless liners, I.Ø: 6,50 mm
  • Seq:
    03
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.