Express Eingabe
Austria
AuftragsverfolgungKontakt
Pasteurpipetten, Glas | Avantor
Pasteurpipetten, Glas
150 mm, -
Artikelnr. 612-3813
Anbieter:   DWK Life Sciences
Pasteurpipetten, Glas
Artikelnr. 612-3813
Anbieter:  DWK Life Sciences
Pasteurpipetten, Glas
Pasteurpipetten, Glas
150 mm, -
Preis
37,50 €
Einloggen für Verfügbarkeit
1 * 250 ST
Product Details & Documents
Pasteur pipette with a narrow tip. The suction tube is designed for cotton plugging.
Dispensing box with 250 pipettes.
Spezifikationen
  • Steril:
    -
  • Länge:
    150 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.