AuftragsverfolgungKontakt
Flasks, round bottom, with three parallel necks | Avantor
Flasks, round bottom, with three parallel necks
1000 ml, 29/32 NS, 14/23 NS
Artikelnr. WITG0.768.001
Anbieter:   witeg Labortechnik
Flasks, round bottom, with three parallel necks
Artikelnr. WITG0.768.001
Anbieter:  witeg Labortechnik
Flasks, round bottom, with three parallel necks
Flasks, round bottom, with three parallel necks
1000 ml, 29/32 NS, 14/23 NS
Preis
66,00 €
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Typical applications: distillation, extraction.
Spezifikationen
  • Kapazität:
    1000 ml
  • Seitenhälse:
    14/23 NS
  • Mittelhals:
    29/32 NS