Deckgläser, rund
Round cover slides, 12 mm, Borosilikatglas
Artikelnr. 631-0666
Anbieter: MARIENFELD
Springe zu:
Product Details & Documents
Round cover glasses made of pure white borosilicate D 263 ™ M glass, Hydrolytic Class I.
- No bubbles or flaws
Suitable for in vitro diagnostic (IVD) applications.
: 100 cover glasses per box with separate lid.
Spezifikationen
- Ø:12 mm
- Bezeichnung:Round cover slides
- Material:Borosilikatglas
- Seq:002