Deckgläser, rund
Round cover slides, 20 mm, Borosilikatglas
Catalog # 631-0670
Supplier: MARIENFELD
Jump to:
Product Details & Documents
Round cover glasses made of pure white borosilicate D 263 ™ M glass, Hydrolytic Class I.
- No bubbles or flaws
Suitable for in vitro diagnostic (IVD) applications.
Packaging: 100 cover glasses per box with separate lid.
Specifications
- Ø:20 mm
- Bezeichnung:Round cover slides
- Material:Borosilikatglas
- Seq:008