AuftragsverfolgungKontakt
Cycloheximid
Cycloheximid
Glass bottle for solids, 1 g
Catalog # BMOL54646-1
Toxic
Health
Environment
Cycloheximid
Catalog # BMOL54646-1
CAS Number:  66-81-9
Cycloheximid
Cycloheximid
Glass bottle for solids, 1 g
CAS Number:  66-81-9
Price
Inquire For Price
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  Naramycin A, Actidione, 3-[2-(3,5-Dimethyl-2-oxocyclohexyl)-2-hydroxyethyl]glutarimide,
  • CAS Number:66-81-9
  • Formula:C₁₅H₂₃NO₄
  • MDL Number:MFCD00082346
  • Molecular Weight:281.35 g/mol
  • Storage Temperature:Raumtemperatur
  • EINECS:200-636-0
  • UN:2811
  • ADR:6.1,I
Specifications
Specifications
  • Packung:
    Glass bottle for solids
  • VE:
    1 g