ContactUsLinkComponent
Sample cup,S.O.E, 32mm diameter by 23mm high for X ray fluorescence 1 * 100 ST
Artikelnr. 710-0132
Lieferant:  CHEMPLEX INDUSTRIES
Sample cup,S.O.E, 32mm diameter by 23mm high for X ray fluorescence 1 * 100 ST
Artikelnr. 710-0132
Lieferant:  CHEMPLEX INDUSTRIES
Frequently Bought Together