Comparator kits, multiparameter for water testing, Comparator 2000+
Sewage and domestic effluent, AF 358, Ammonia: 0 - 1 mg/l Nl
Chlorine: 0,1 - 1 mg/l Cl₂
Chlorine: 1 - 10 mg/l Cl₂
Nitrite: 0,05 - 0,5 mg/l N
Permanganate (BOD): 0 - 60 mg/l
pH value: 4 - 8; 8 - 9,6
Sulphide: 0 - 0,5 mg/l S
Chlorine: 0,1 - 1 mg/l Cl₂
Chlorine: 1 - 10 mg/l Cl₂
Nitrite: 0,05 - 0,5 mg/l N
Permanganate (BOD): 0 - 60 mg/l
pH value: 4 - 8; 8 - 9,6
Sulphide: 0 - 0,5 mg/l S
705-0362
Lovibond® Comparator system 2000+ is an extremely versatile, modular system for testing water. The battery operated portable lighting unit can be integrated to give standardised lighting conditions to enhance reproducibility.
- Compact and portable
- Simple to use yet uncompromising in terms of precision and reproducibility of results
: Comparator test kits are supplied as a complete system in a sturdy plastic case. Together with the Comparator 2000+ and test discs, each kit includes all the necessary cells, accessories and Lovibond® tablet reagents (for 100 measurements). The AF368 is not supplied with reagents.
- Messbereich:Ammonia: 0 - 1 mg/l Nl
Chlorine: 0,1 - 1 mg/l Cl₂
Chlorine: 1 - 10 mg/l Cl₂
Nitrite: 0,05 - 0,5 mg/l N
Permanganate (BOD): 0 - 60 mg/l
pH value: 4 - 8; 8 - 9,6
Sulphide: 0 - 0,5 mg/l S - Bezeichnung:Sewage and domestic effluent
- Seq:03
- Typ:AF 358