Surgical scissors, type Mayo
Gekrümmt, abgeschrägte Klingen, 165 mm
Catalog # MURR1882/10
Supplier: MURRAY S
Jump to:
Product Details & Documents
These scissors can be used for a variety of applications in medical areas, laboratories and microscopy.
- Straight or curved
- Bevelled blades
- Blunt
Specifications
- Typ:Gekrümmt, abgeschrägte Klingen
- Länge:165 mm
- Seq:03
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.