Biopsy specimen processing, CellSafe™
Biowrap 60x60 mm, white
Catalog # 720-2355
Supplier: CELLPATH
Jump to:
Product Details & Documents
Consists of two hinged interlocking frames with an integral nylon mesh. This very fine 200 µm mesh ensures a high degree of protection for specimens.
- No tissue damage
- Low reagent carry-over
The CellSafe™ fits in every embedding cassette.
Specifications
- Bezeichnung:Biowrap 60x60 mm, white