Loops, disposable, Microloops
Impfösen in Peel-Off-Verpackung, 10 µl, Rot
Supplier: MEDICAL WIRE EQUIPMENT
Jump to:
Product Details & Documents
Sterile, disposable inoculating loops.
- Colour coded plastic
- Moderately flexible
Specifications
- Kapazität:10 µl
- Farbe:Rot
- Bezeichnung:Impfösen in Peel-Off-Verpackung