Express Eingabe
Austria
AuftragsverfolgungKontakt
Cover slips for microscope slides | Avantor

Cover slips for microscope slides

Cover slip for microscope slide, - mm, 0,15 - 0,25 mm, 25,4 mm, Quartz
Artikelnr. 43211.KG
Cover slips for microscope slides
Artikelnr. 43211.KG
Cover slips for microscope slides

Cover slip for microscope slide, - mm, 0,15 - 0,25 mm, 25,4 mm, Quartz

Preis
74,80 €
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents

These cover slips made of fused quartz, have many properties, due to the fusing of naturally occuring crystalline silica.

  • High chemical purity and corrosion resistance
  • Low thermal expansion coefficient
  • Extreme hardness

Excellent electrical insulation and optical transmission from ultraviolet to infrared are suitable for analytical purposes.

Dokumente

Zertifikate
Suche nach Chargennummer
Spezifikationen
  • Ø:
    25,4 mm
  • Bezeichnung:
    Cover slip for microscope slide
  • L×B:
    - mm
  • Material:
    Quartz
  • Seq:
    02
  • Stärke:
    0,15 - 0,25 mm