Dimethyldisulfid ≥99%
Glasflasche für Feststoffe, 250 g
Anbieter: Thermo Fisher Scientific
Springe zu:
Product Details & Documents
Purity: 99% CAS No.: 624-92-0 Molecular Formula: C2H6S2 Formula Weight: 94.20 Flash Point: 10°(50°F) Refractive Index: 1.5253 Odour: Stench Melting Point: -85° Boiling Point: 108-109° Beilstein Reference: 1730824 UN No.: 2381 MDL No.: MFCD00008561
Synonyme: Dimethyl disulphide, Methyl disulphide
- CAS Nummer:624-92-0
- Formel:C₂H₆S₂
- MDL-Nummer:MFCD00008561
- Molekulargewicht:94.2 g/mol
- Siedepunkt:108…109 °C
- Schmelzpunkt:–85 °C
- Dichte:1,046
- Blitzpunkt:10 °C (50 °F)
- Lagertemperatur:Raumtemperatur
- EINECS:210-871-0
- UN:2381
- ADR:3,II
Documents
Zertifikate
Suche nach ChargennummerALFAL09679.30_TDS_EN
Product specification
Organic Building Blocks and Intermediates
Application Notes
Spezifikationen
Spezifikationen
- Packung:Glasflasche für Feststoffe
- VE:250 g
Spezifikationen
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



