Express Eingabe
Austria
AuftragsverfolgungKontakt
Büchner-Trichter

Büchner-Trichter

95 mm, 500 ml, 3
Catalog # 511-0246
Supplier:   SciLabware
Büchner-Trichter
Catalog # 511-0246
Supplier:  SciLabware
Büchner-Trichter
Büchner-Trichter

95 mm, 500 ml, 3

Price
537,00 €
Log in for availability
1 * 5 ST
Product Details & Documents

PYREX®, borosilicate glass, with sintered glass disc.

  • No filter papers required
  • Highly resistant to most corrosive reagents including ammonia and concentrated sulphuric acid
  • Very good temperature resistance

For convenient filtration of solutions.

Specifications
  • Platte Ø:
    95 mm
  • Porosität:
    3
  • Kapazität:
    500 ml
  • Seq:
    40
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.