Express Eingabe
Austria
AuftragsverfolgungKontakt
Büchner-Trichter

Büchner-Trichter

40 mm, 50 ml, 2
Catalog # BIBB3630/04
Supplier:   SciLabware
Büchner-Trichter
Catalog # BIBB3630/04
Supplier:  SciLabware
Büchner-Trichter
Büchner-Trichter

40 mm, 50 ml, 2

Price
222,00 €
Log in for availability
1 * 5 ST
Product Details & Documents

PYREX®, borosilicate glass, with sintered glass disc.

  • No filter papers required
  • Highly resistant to most corrosive reagents including ammonia and concentrated sulphuric acid
  • Very good temperature resistance

For convenient filtration of solutions.

Specifications
  • Platte Ø:
    40 mm
  • Porosität:
    2
  • Kapazität:
    50 ml
  • Seq:
    14