Express Eingabe
Austria
AuftragsverfolgungKontakt
 

Büchner-Trichter

30 mm, 30 ml, 5
Catalog # BIBB3620/10
Supplier:   SciLabware
Büchner-Trichter
Catalog # BIBB3620/10
Supplier:  SciLabware
Büchner-Trichter
Büchner-Trichter

30 mm, 30 ml, 5

Price
953,00 €
Log in for availability
1 * 20 ST
Product Details & Documents

PYREX®, borosilicate glass, with sintered glass disc.

  • No filter papers required
  • Highly resistant to most corrosive reagents including ammonia and concentrated sulphuric acid
  • Very good temperature resistance

For convenient filtration of solutions.

Specifications
  • Platte Ø:
    30 mm
  • Porosität:
    5
  • Kapazität:
    30 ml
  • Seq:
    08