Büchner-Trichter
30 mm, 30 ml, 3
Catalog # BIBB3620/06
Supplier: SciLabware
Jump to:
Product Details & Documents
PYREX®, borosilicate glass, with sintered glass disc.
- No filter papers required
- Highly resistant to most corrosive reagents including ammonia and concentrated sulphuric acid
- Very good temperature resistance
For convenient filtration of solutions.
Specifications
- Platte Ø:30 mm
- Porosität:3
- Kapazität:30 ml
- Seq:04



