Express Eingabe
Austria
AuftragsverfolgungKontakt
 

Büchner-Trichter

30 mm, 30 ml, 2
  BIBB3620/04
 :   SciLabware
Büchner-Trichter
  BIBB3620/04
Büchner-Trichter
Büchner-Trichter

30 mm, 30 ml, 2

 
232,00 €
   
1 * 5 ST
 

PYREX®, borosilicate glass, with sintered glass disc.

  • No filter papers required
  • Highly resistant to most corrosive reagents including ammonia and concentrated sulphuric acid
  • Very good temperature resistance

For convenient filtration of solutions.

 
  • Platte Ø:
    30 mm
  • Porosität:
    2
  • Kapazität:
    30 ml
  • Seq:
    02