Express Eingabe
Austria
AuftragsverfolgungKontakt
 

Büchner-Trichter

30 mm, 30 ml, 1
  BIBB3620/02
 :   SciLabware
Büchner-Trichter
  BIBB3620/02
Büchner-Trichter
Büchner-Trichter

30 mm, 30 ml, 1

 
232,00 €
   
1 * 5 ST
 

PYREX®, borosilicate glass, with sintered glass disc.

  • No filter papers required
  • Highly resistant to most corrosive reagents including ammonia and concentrated sulphuric acid
  • Very good temperature resistance

For convenient filtration of solutions.

 
  • Platte Ø:
    30 mm
  • Porosität:
    1
  • Kapazität:
    30 ml
  • Seq:
    01
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.