Desiccators, PYREX®
Rotaflo® stopcock with 24/29 joint for vacuum tight seal
Artikelnr. BIBB1612/03
Anbieter: SciLabware
Springe zu:
Product Details & Documents
Ideal for vacuum and non vacuum applications.
- Ground glass flanges
- Borosilicate glass base and lid
- Plates positioned on an internal ledge within the base
Spezifikationen
- Bezeichnung:Rotaflo® stopcock with 24/29 joint for vacuum tight seal
- Cat. No.:BIBB1612/03