AuftragsverfolgungKontakt
Spherical ground joints, Quickfit® | Avantor
Spherical ground joints, Quickfit®
S 35, -
Artikelnr. BIBBMS35UB
Anbieter:   SciLabware
Spherical ground joints, Quickfit®
Artikelnr. BIBBMS35UB
Anbieter:  SciLabware
Spherical ground joints, Quickfit®
Spherical ground joints, Quickfit®
S 35, -
Preis
152,00 €
Einloggen für Verfügbarkeit
1 * 5 ST
Product Details & Documents
Made of PYREX®, borosilicate glass.
  • Excellent sealing properties
  • Minimal requirement for joint lubrication
  • Ideal for use as flexible connectors
For the Glassblower who demands the highest quality joint.
Length: 100 mm
Spezifikationen
  • Kugelschliff-Kugel:
    S 35
  • Kugelschliff-Schale:
    -