Pulvertrichter
100 mm, 26 mm, 25 mm
Artikelnr. SEMA0211
Anbieter: SEMADENI
Springe zu:
Product Details & Documents
Funnels, made of PP, translucent. Used for transferring powders. Also suitable for liquids.
- Parallel stem minimises powder bridging
- Very good chemical and temperature resistance
- Autoclavable
Spezifikationen
- Trichter A-Ø:100 mm
- Stiel A-Ø:26 mm
- Stiel Länge:25 mm



