Natriumborhydrid
Glass bottle for solids, 10 g
MAYBSB01509.10
Marketsource
: Sodium tetrahydridoborate, Sodium tetrahydroborate
- :16940-66-2
- :NaBH₄
- :MFCD00003518
- :37.83 g/mol
- :13,08664
- :400 °C
- :1,07 g/cm³ (20 °C)
- :69 °C
- :Raumtemperatur
- :241-004-4
- :1426
- :4.3,I
- Packung:Glass bottle for solids
- VE:10 g