Benzofuran
Glass bottle for solids, 10 g
MAYBSB00762.10
Marketsource
: 2,3-Benzofuran, Benzo[b]furan, Benzo(b)furan, 1-Benzofuran, Coumarone
- :271-89-6
- :C₈H₆O
- :MFCD00005847
- :118.14 g/mol
- :13,01089
- :173 °C (1013 hPa)
- :< –18 °C
- :1,1 g/cm³ (20 °C)
- :50 °C
- :Density: 1.1 g/cm³ (20 °C)
- :205-982-6
- Packung:Glass bottle for solids
- VE:10 g