4'-Hydroxy-3'-methoxyacetophenon
Glass bottle for solids, 250 mg
CHMPNRB05256.250
: CHEMPUR
: 4-Hydroxy-3-methoxy-acetophenone, 1-(4-Hydroxy-3-methoxyphenyl)ethanone, Acetovanillone, Apocynin
- :498-02-2
- :C₉H₁₀O₃
- :MFCD00008747
- :166.18 g/mol
- :13,00746
- :295 °C (1013 hPa)
- :115 °C
- :207-854-5
- Packung:Glass bottle for solids
- VE:250 mg