AuftragsverfolgungKontakt
4'-Hydroxy-3'-methoxyacetophenon
Glass bottle for solids, 250 mg
  CHMPNRB05256.250
 :   CHEMPUR
4'-Hydroxy-3'-methoxyacetophenon
  CHMPNRB05256.250
 :  CHEMPUR
 :  498-02-2
4'-Hydroxy-3'-methoxyacetophenon
Glass bottle for solids, 250 mg
 :  498-02-2
1 * 250 mg
This product has been blocked by your organization. Please contact your purchasing department for more information
 :  4-Hydroxy-3-methoxy-acetophenone, 1-(4-Hydroxy-3-methoxyphenyl)ethanone, Acetovanillone, Apocynin
  •  :498-02-2
  •  :C₉H₁₀O₃
  •  :MFCD00008747
  •  :166.18 g/mol
  •  :13,00746
  •  :295 °C (1013 hPa)
  •  :115 °C
  •  :207-854-5
  • Packung:
    Glass bottle for solids
  • VE:
    250 mg