ω-Brom-4-fluoracetophenon
Glass bottle for solids, 1 g
Artikelnr. MAYBBTB05530.1
Marketsource
Anbieter: Thermo Scientific
ω-Brom-4-fluoracetophenon
Artikelnr. MAYBBTB05530.1
Anbieter: Thermo Scientific
Marketsource
CAS Nummer: 403-29-2
Springe zu:
Product Details & Documents
Synonyme: Bromomethyl p-fluorophenyl ketone, α-Bromo-4-fluoroacetophenone, 2-Bromo-1-(4-fluorophenyl)ethan-1-one, 2-Bromo-4'-fluoroacetophenone, 4-Fluorophenacyl bromide, 2-Bromo-1-(4-fluorophenyl)ethanone
- CAS Nummer:403-29-2
- Formel:C₈H₆BrFO
- MDL-Nummer:MFCD00040830
- Molekulargewicht:217.04 g/mol
- Siedepunkt:82 °C (0,4 mmHg)
- Schmelzpunkt:49 °C
- EINECS:206-955-1
Spezifikationen
Spezifikationen
- Packung:Glass bottle for solids
- VE:1 g
Spezifikationen
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



