Express Eingabe
Austria
AuftragsverfolgungKontakt
Weighing boats
Weighing boats
10×4×4,5 mm
Catalog # LUDI176982816
Supplier:   LUDI
Weighing boats
Catalog # LUDI176982816
Supplier:  LUDI
Weighing boats
10×4×4,5 mm
Price
64,60 €
Log in for availability
1 * 250 ST
Product Details & Documents
Made of aluminium.
  • Lightweight
  • Good chemical resistance
  • Good temperature resistance
Specifications
  • Abmessungen:
    10×4×4,5 mm
  • Seq:
    04
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.